Learn More
Quality Biological Inc 50 mL Reagent Reservoir - Polystyrene, V-Shaped, Bulk Pack, White, Sterile, 5/PK, 200/CS
SDP
Catalog No. 50147431
Encompass_Preferred
50 mL Reagent Reservoir - Polystyrene, Bulk, White, Sterile
Packaged 5/Bag, 200/CS
• Reagent Reservoirs are designed to assist in repetitive liquid transfer
• White color enhances solution visibility
• Angled design promotes funneling of liquid to maximize retrieval
Catalog No. 50-147-431 Supplier Quality Biological Inc Supplier No. P886229291
50 mL Reagent Reservoir - Polystyrene, Bulk, White, Sterile
Packaged 5/Bag, 200/CS
• Reagent Reservoirs are designed to assist in repetitive liquid transfer
• White color enhances solution visibility
• Angled design promotes funneling of liquid to maximize retrieval