Learn More
Bellco Glass, Inc. 50mL Reagent Reservoir
SDP
Catalog No. 502329856
Encompass_Preferred
DescriptionReagent Reservoirs are designed to assist in repetitive liquid transferWhite color enhances solution visibilityAngled design promotes funneling of liquid to maximize retrieval Pack Description5/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolystyreneMaximum Volume50mLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-232-9856 Supplier Bellco Glass, Inc. Supplier No. 229291000
DescriptionReagent Reservoirs are designed to assist in repetitive liquid transferWhite color enhances solution visibilityAngled design promotes funneling of liquid to maximize retrieval Pack Description5/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolystyreneMaximum Volume50mLGraduatedNoRNase/DNase FreeYes