Learn More
Quality Biological Inc 50 mL Reagent Reservoir - Polystyrene, V-Shaped, Individual Pack, White, Sterile, 1/PK, 80/CS
SDP
Catalog No. 50147430
Encompass_Preferred
50 mL Reagent Reservoir - Polystyrene, V-Shaped, Individual Pack, White, Sterile
Packaged 1/PK, 80/CS
• Reagent Reservoirs are designed to assist in repetitive liquid transfer
• White color enhances solution visibility
• Angled design promotes funneling of liquid to maximize retrieval
Catalog No. 50-147-430 Supplier Quality Biological Inc Supplier No. P886229290
50 mL Reagent Reservoir - Polystyrene, V-Shaped, Individual Pack, White, Sterile
Packaged 1/PK, 80/CS
• Reagent Reservoirs are designed to assist in repetitive liquid transfer
• White color enhances solution visibility
• Angled design promotes funneling of liquid to maximize retrieval