Learn More
Sarstedt Inc 50ml Screen Printed Screw Cap Tube, Conical Bottom, Polypropylene, Non-Sterile, 300/Case
Catalog No. NC0039591 Shop All Sarstedt Inc. Products
Encompass_Preferred
50ml Screen Printed Screw Cap Tube 28x114mm Conical Bottom Skirted Polypropylene Graduations and Writing Block Neutral Cap. Non-sterile.
Catalog No. NC0039591 Supplier Sarstedt Inc Supplier No. 62.559.101
50ml Screen Printed Screw Cap Tube 28x114mm Conical Bottom Skirted Polypropylene Graduations and Writing Block Neutral Cap. Non-sterile.