Learn More
Sarstedt Inc 50mL Screen Printed Screw Cap Tube, Conical Bottom, Polypropylene, 300/Case
Catalog No. NC9174817 Shop All Sarstedt Inc. Products
Encompass_Preferred
50mL Screen Printed Screw Cap Tube 28x114mm Conical Bottom Polypropylene Graduations and Writing Block Bulk Pack Red Cap
Catalog No. NC9174817 Supplier Sarstedt Inc Supplier No. 62.548.101
50mL Screen Printed Screw Cap Tube 28x114mm Conical Bottom Polypropylene Graduations and Writing Block Bulk Pack Red Cap