Learn More
Sarstedt Inc Screw Cap Tube 50ml, 28X114mm, PP, conical base with skirted bottom, 25/bag, 12 bags/case, 300 total
Supplier:  Sarstedt Inc 62.559.205
Encompass_Preferred
Screw cap tube, 50 ml, 114 x 28 mm, conical base with skirted bottom, PP, with printed writing space and graduation, with assembled cap, 25 pcs./bag , 12 bags/case, 300 total
Catalog No. NC9874179