Learn More
Quality Biological Inc 50 mL Self-Standing Bio-Reaction Tube - Sterile, Bulk Pack, 10/Resealable Bag, 100/CS
SDP
Catalog No. 50147458
Encompass_Preferred
50 mL Self-Standing Bio-Reaction Tube, Sterile, Bulk Pack
Packaged 10/Resealable Bag, 100/CS
• Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture
• Ideal for biological reaction applications and free gas exchange
• Aseptic conditions maintained through 0.22 μm hydrophobic vented membrane
• Useful in the screening and optimization processes of suspension cells
• Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously
• Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization
• Vent control labels for 50mL Bio-Reaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange
Catalog No. 50-147-458 Supplier Quality Biological Inc Supplier No. P886229479
50 mL Self-Standing Bio-Reaction Tube, Sterile, Bulk Pack
Packaged 10/Resealable Bag, 100/CS
• Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture
• Ideal for biological reaction applications and free gas exchange
• Aseptic conditions maintained through 0.22 μm hydrophobic vented membrane
• Useful in the screening and optimization processes of suspension cells
• Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously
• Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization
• Vent control labels for 50mL Bio-Reaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange