Learn More
CELLTREAT Scientific Products 50mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Catalog No. 50969838
Encompass_Preferred
CELLTREAT Scientific Products 50mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Negative graduations of -10mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic wrappers. 25/bag, 100/case.
Catalog No. 50-969-838 Supplier CELLTREAT Scientific Products Supplier No. 229030B
CELLTREAT Scientific Products 50mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Negative graduations of -10mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic wrappers. 25/bag, 100/case.