Learn More
CELLTREAT Scientific Products 50mL Serological Pipet, Paper/Plastic Wrapped, Sterile
SDP
Catalog No. NC1772272
Encompass_Preferred
CELLTREAT Scientific Products 50mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.
Catalog No. NC1772272 Supplier CELLTREAT Scientific Products Supplier No. 229031B
CELLTREAT Scientific Products 50mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.