Learn More
CELLTREAT Scientific Products 50mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
SDP
Supplier:  CELLTREAT Scientific Products 229230B
Encompass_Preferred
CELLTREAT Scientific Products 50mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 30 packed/bag, 90/case.
Catalog No. 50-202-112