Learn More
Bellco Glass, Inc. 50ml Serological Pipette, Individually Wrapped sterile 100/cs, 600/cs
SDP
Catalog No. 502330259
Encompass_Preferred
50ml Serological Pipette with 6.0ml negative graduation, 5/10 ml graduations, Individually Plastic-plastic Wrapped sterile, 25/bag, 100/cs, 600/cs. Plastic/plastic packaging permits peel-off function that provides user-friendly easy opening and aseptic pipette removal, reduced contamination risk and particulate-free opening. Fine line graduation printing contributes to accuracy of at least +/- 2%. Reduced contamination risk and particulate-free openingFine line graduation printing contributes to accuracy of at least +/- 2%Negative graduations provide increased volume versatilityReverse (ascending and descending) graduations on 5, 10, 25 and 50 mL pipettesSynthetic filter and color-coded according to international conventionLot number and best before date on each individual wrap
Catalog No. 50-233-0259 Supplier Bellco Glass, Inc. Supplier No. 451400248
50ml Serological Pipette with 6.0ml negative graduation, 5/10 ml graduations, Individually Plastic-plastic Wrapped sterile, 25/bag, 100/cs, 600/cs. Plastic/plastic packaging permits peel-off function that provides user-friendly easy opening and aseptic pipette removal, reduced contamination risk and particulate-free opening. Fine line graduation printing contributes to accuracy of at least +/- 2%. Reduced contamination risk and particulate-free openingFine line graduation printing contributes to accuracy of at least +/- 2%Negative graduations provide increased volume versatilityReverse (ascending and descending) graduations on 5, 10, 25 and 50 mL pipettesSynthetic filter and color-coded according to international conventionLot number and best before date on each individual wrap