Learn More
Neuromics, Inc Strep-Tactin 4Flow High Capacity Resin, 50 ml suspension, protein purification, Strep-tag II and Twin-Strep-tag proteins, 4% cross-linked agarose beads, Strep-Tactin ligand, >95% purity
Catalog No. NC3664194
Encompass
Strep-Tactin 4Flow High Capacity Resin, 50 ml suspension, protein purification, Strep-tag II and Twin-Strep-tag proteins, 4% cross-linked agarose beads, Strep-Tactin ligand, >95% purity
Catalog No. NC3664194 Supplier Neuromics, Inc Supplier No. 21250025
Strep-Tactin 4Flow High Capacity Resin, 50 ml suspension, protein purification, Strep-tag II and Twin-Strep-tag proteins, 4% cross-linked agarose beads, Strep-Tactin ligand, >95% purity