Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube - Self-Standing, Bag, Sterile
SDP
Catalog No. 50202008
Encompass_Preferred
CELLTREAT Scientific Products 50mL Self-standing Centrifuge Tube. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Self-standing base for use without a rack. RNase/DNase free and non-pyrogenic. Max force (RFC) g 6000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 500/case.
Catalog No. 50-202-008 Supplier CELLTREAT Scientific Products Supplier No. 229430
CELLTREAT Scientific Products 50mL Self-standing Centrifuge Tube. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Self-standing base for use without a rack. RNase/DNase free and non-pyrogenic. Max force (RFC) g 6000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 500/case.