Learn More
Savillex Corporation 50 mL Standard Tube, Conical Interior
SDP
Catalog No. NC1612141
Encompass
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). PFA non‑breakable. Threaded top for long term storage. Fits most standard centrifuge rotors. RCF up to 5,000 g.
Catalog No. NC1612141 Supplier Savillex Corporation Supplier No. 21005030
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). PFA non‑breakable. Threaded top for long term storage. Fits most standard centrifuge rotors. RCF up to 5,000 g.