Learn More
Chemglass Life Sciences 50mL Storage Flask, #15 O-Ring jt, Airfree, Schlenk, 0-4mm Valve, 120mm OAH
SDP
Catalog No. AF009301
Encompass_Preferred
50mL Storage Flask, #15 O-Ring jt, Airfree, Schlenk, 0-4mm Valve, 120mm OAH. Flask has a #15 o-ring joint on the sidearm and a Chem-Vac™ Chem-Cap valve to control gas or liquid flow into the vessel. All sizes are of heavy wall construction, and are used with AF-0060 and AF-0061 manifolds or any other Airfree component with a #15 o-ring joint. Supplied complete with one #116 Viton o-ring.
Catalog No. AF009301 Supplier Chemglass Life Sciences Supplier No. AF009301
50mL Storage Flask, #15 O-Ring jt, Airfree, Schlenk, 0-4mm Valve, 120mm OAH. Flask has a #15 o-ring joint on the sidearm and a Chem-Vac™ Chem-Cap valve to control gas or liquid flow into the vessel. All sizes are of heavy wall construction, and are used with AF-0060 and AF-0061 manifolds or any other Airfree component with a #15 o-ring joint. Supplied complete with one #116 Viton o-ring.