Learn More
Chemglass Life Sciences 50mL Storage Vessel, Cajon, Airfree, Schlenk, 0-4mm Valve, 190mm OAH
SDP
Catalog No. AF009602
Encompass_Preferred
50mL Storage Vessel, Cajon, Airfree, Schlenk, 0-4mm Valve, 190mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.
Catalog No. AF009602 Supplier Chemglass Life Sciences Supplier No. AF009602
50mL Storage Vessel, Cajon, Airfree, Schlenk, 0-4mm Valve, 190mm OAH. Storage vessel sidearm has a 9mm OD tubulation for use with 3/8" Cajon connections. Chem-Vac™ Chem-Cap valve is used to control gas or liquid flow into the vessel.