Learn More
Chemglass Life Sciences Sublimation Apparatus, 50mL, Complete
SDP
Catalog No. 501531535
Encompass_Preferred
Sublimation Apparatus, 50mL, Complete. Two-piece sublimer consisting of an upper water cooled condenser and lower flat bottom flask. Sublimate is collected on the inner wall of the condenser. The two components are assembled by a grease-free o-ring seal which permits the components to be gently separated for maximum product recovery. When condenser is inverted, the top vacuum connection serves as a funnel for convenient product removal. Complete apparatus consists of a condenser, flask, Viton o-ring and CG-150 Stainless Steel clamp.
Catalog No. 50-153-1535 Supplier Chemglass Life Sciences Supplier No. CG303602
Sublimation Apparatus, 50mL, Complete. Two-piece sublimer consisting of an upper water cooled condenser and lower flat bottom flask. Sublimate is collected on the inner wall of the condenser. The two components are assembled by a grease-free o-ring seal which permits the components to be gently separated for maximum product recovery. When condenser is inverted, the top vacuum connection serves as a funnel for convenient product removal. Complete apparatus consists of a condenser, flask, Viton o-ring and CG-150 Stainless Steel clamp.