Learn More
CELLTREAT Scientific Products 50mL Suspension Culture Flask - Vent Cap, Sterile
SDP
Catalog No. 50202084
Encompass_Preferred
CELLTREAT Scientific Products 50mL Suspension Culture Flask - Non-treated. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Vent caps feature 0.22¿m hydrophobic membrane. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 200/case.
Catalog No. 50-202-084 Supplier CELLTREAT Scientific Products Supplier No. 229510
CELLTREAT Scientific Products 50mL Suspension Culture Flask - Non-treated. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Vent caps feature 0.22¿m hydrophobic membrane. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 200/case.