Learn More
Research Products International Corp Scienceware 50 ml Tube Holder, Grip Style, Blue, 5 per Case
SDP
Catalog No. 50998383
Encompass
This unique design tube base holds any 50ml conical tube and leaves both hands free for pipetting or sampling procedures. The grip style holder remains attached when lifting the tube from the bench top. Tube can be removed by holding base and pulling tube out. The non-grip base securely holds the tube in place and allows tube to be lifted freely from holder.
Catalog No. 50-998-383 Supplier Research Products International Corp Supplier No. 140215
This unique design tube base holds any 50ml conical tube and leaves both hands free for pipetting or sampling procedures. The grip style holder remains attached when lifting the tube from the bench top. Tube can be removed by holding base and pulling tube out. The non-grip base securely holds the tube in place and allows tube to be lifted freely from holder.