Learn More
Waters Corp Tube Rack, 50 mL, 55-Position, Domino
Catalog No. NC3597586 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rack domino is designed to accommodate a large format rack capable of holding up to 55 tubes. It features a 3x2 format, occupying the space of six standard dominos on an Andrew+ robot workbench. This design allows for seamless automation of laboratory workflows with the Andrew+ pipetting robot.
  • Machine-readable consumable holder interfaces with Andrew+ pipetting robot.
  • Facilitates seamless automation of laboratory workflows.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Essential for Andrew+ automation configurations.
  • Designed to hold a very large format rack capable of holding up to 55 tubes.
  • Features a 3x2 format on the Andrew+ robot workbench.
  • Occupies three positions horizontally and two positions vertically.
  • Equivalent to the space taken by six standard dominos.
Catalog No. NC3597586 Supplier Waters Corp Supplier No. 186010514
Add to Cart
Add to Cart
Request a Quote
This rack domino is designed to accommodate a large format rack capable of holding up to 55 tubes. It features a 3x2 format, occupying the space of six standard dominos on an Andrew+ robot workbench. This design allows for seamless automation of laboratory workflows with the Andrew+ pipetting robot.
  • Machine-readable consumable holder interfaces with Andrew+ pipetting robot.
  • Facilitates seamless automation of laboratory workflows.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
  • Essential for Andrew+ automation configurations.
  • Designed to hold a very large format rack capable of holding up to 55 tubes.
  • Features a 3x2 format on the Andrew+ robot workbench.
  • Occupies three positions horizontally and two positions vertically.
  • Equivalent to the space taken by six standard dominos.