Learn More
Waters Corp Tube Magnet, 50 mL, DNA Purification, Magnetic Bead Separation
Catalog No. 502391617 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This device offers a straightforward and dependable method for purifying genomic, plasmid, and mitochondrial DNA using magnetic beads.
  • Features an automated moving shelf that precisely positions magnets close to the bottom sides of consumables.
  • Optimizes bead pelleting for efficient purification.
  • Suitable for genomic, plasmid, and mitochondrial DNA purification.
Catalog No. 50-239-1617 Supplier Waters Corp Supplier No. 176004851
Add to Cart
Add to Cart
Request a Quote
This device offers a straightforward and dependable method for purifying genomic, plasmid, and mitochondrial DNA using magnetic beads.
  • Features an automated moving shelf that precisely positions magnets close to the bottom sides of consumables.
  • Optimizes bead pelleting for efficient purification.
  • Suitable for genomic, plasmid, and mitochondrial DNA purification.