Learn More
Ace Glass, Inc. 50ML TUBE ONLY 50ML TUBE ONLY
SDP
Catalog No. 50974572
Encompass_Preferred
50mL glass tapered tube with a pouring lip, for use with a glass pestle (50974573) as a tissue grinder. The conical area of the tube is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1-0.15mm clearance. Capacity represents volume with pestle inserted.
This item is not returnable. View return policy
Catalog No. 50-974-572 Supplier Ace Glass, Inc. Supplier No. 832528
50mL glass tapered tube with a pouring lip, for use with a glass pestle (50974573) as a tissue grinder. The conical area of the tube is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1-0.15mm clearance. Capacity represents volume with pestle inserted.