Learn More
ALPAQUA ENGINEERING LLC 50ML TUBE RACK (1EA)
Catalog No. NC0561871
Encompass
The 6 position 50ml tube rack from Alpaqua is an SBS compliant deck accessory allowing use on a variety of robotic platforms. It is designed to accommodate tubes up to 30mm in diameter.
Catalog No. NC0561871 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000290
The 6 position 50ml tube rack from Alpaqua is an SBS compliant deck accessory allowing use on a variety of robotic platforms. It is designed to accommodate tubes up to 30mm in diameter.