Learn More
Andwin Scientific Rack for 50 mL Tubes, Assorted, 5/cs
SDP
Catalog No. 501126045
Encompass_Preferred
These racks are designed to hold centrifuge tubes and replace foam racks. Racks also feature an interlocking design that allows users to connect more than one rack and create a handy work station. These racks are designed for Labcon centrifuge tubes, other tube brands may not fit. Unlike foam racks, they are autoclavable and will not fall apart with repeated use. They can be frozen, and will stack together to minimize storage space.
Catalog No. 50-112-6045 Supplier Andwin Scientific Supplier No. 89079512
These racks are designed to hold centrifuge tubes and replace foam racks. Racks also feature an interlocking design that allows users to connect more than one rack and create a handy work station. These racks are designed for Labcon centrifuge tubes, other tube brands may not fit. Unlike foam racks, they are autoclavable and will not fall apart with repeated use. They can be frozen, and will stack together to minimize storage space.