Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube Filter, 0.22m PES Filter, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229710
Encompass_Preferred
CELLTREAT Scientific Prdoucts 50mL Centrifuge, 0.22¿m PES Filter System. Eliminate a sample transfer step by filtering directly into a 50mL tube. Polyethersulfone (PES) membrane for high flow and low binding. One Centrifuge Tube Stand included with each case of 50mL Centrifuge Tube Filter Systems. Centrifuge tube Stands are manufactured from polypropylene and are autoclavable, reusable and recyclable. Centrifuge Tube Stand will hold one 50mL tube, three 15mL tubes and three microcentrifuge tubes (1.5mL). Membrane diameter 50mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 12/case.
Catalog No. 50-607-519