Learn More
Syringa Lab Supplies LLC 50ML UNCUT-CAPS ONLY- PK/25
Catalog No. NC2540823
Encompass
These simple yet effective lids will assist in keeping airborne contaminates out of your samples. The caps are the Jet Biofil brand and work with many popular brands of centrifuge tubes.1.6 mm thick, 50 durometer, FDA grade silicone septumAutoclavable to 121°CCAPS ONLYFits most 50 mL centrifuge tubesJet Biofil brand caps
Catalog No. NC2540823 Supplier Syringa Lab Supplies LLC Supplier No. 1J920
These simple yet effective lids will assist in keeping airborne contaminates out of your samples. The caps are the Jet Biofil brand and work with many popular brands of centrifuge tubes.1.6 mm thick, 50 durometer, FDA grade silicone septumAutoclavable to 121°CCAPS ONLYFits most 50 mL centrifuge tubesJet Biofil brand caps