Learn More
Bellco Glass, Inc. 50mL Wobble-not Serological Pipet, Individually Wrapped, Sterile
SDP
Catalog No. 502330032
Encompass_Preferred
Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end, requiring less insertion forceWobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce drippingCompatible with any pipet controllerUltra-clear markings and color-coded stripes for quick volume identification25 & 50mL pipets feature negative graduations to -10mLPremium polymer fiber barrier plugAccuracy ± 2% at full volume Pack Description25 Individually Wrapped/BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorPurplePrimary MaterialPolystyreneVolume50mLGraduatedYesGraduation Increments (Subdivisions)5/10mLNegative Graduations10mLRNase/DNase FreeYes
Catalog No. 50-233-0032 Supplier Bellco Glass, Inc. Supplier No. 229094B00
Wobble-not serological pipets have a patented innovative 2-tier step design at the plug end vs. traditional pipets that have a straight cylindrical-shaped plug end, requiring less insertion forceWobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce drippingCompatible with any pipet controllerUltra-clear markings and color-coded stripes for quick volume identification25 & 50mL pipets feature negative graduations to -10mLPremium polymer fiber barrier plugAccuracy ± 2% at full volume Pack Description25 Individually Wrapped/BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorPurplePrimary MaterialPolystyreneVolume50mLGraduatedYesGraduation Increments (Subdivisions)5/10mLNegative Graduations10mLRNase/DNase FreeYes