Learn More
CELLTREAT Scientific Products 50mL Wobble-not Serological Pipet, Individually Wrapped, Paper/Plastic, Bag, Sterile
SDP
Catalog No. NC2206332
Encompass_Preferred
No Wobble. No Drip. Wobble-not serological pipets require less insertion force due to the patented, innovative 2-tier step design. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce dripping. Ultra-clear markings and color-coded stripes for quick volume identification. Accuracy ± 2% at full volume. Ideal liquid handling partner when paired with ali-Q™ 2 Pipet Controller.
Catalog No. NC2206332 Supplier CELLTREAT Scientific Products Supplier No. 229094B
No Wobble. No Drip. Wobble-not serological pipets require less insertion force due to the patented, innovative 2-tier step design. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controller’s nozzle, to increase stability and reduce dripping. Ultra-clear markings and color-coded stripes for quick volume identification. Accuracy ± 2% at full volume. Ideal liquid handling partner when paired with ali-Q™ 2 Pipet Controller.