Learn More
Ace Glass, Inc. 50mm 10 microliter, 26 gauge, removable needle syringe
SDP
Catalog No. 50972635
Encompass
10 microliter syringe with 50mm 26 gauge removable needle for precise liquid delivery.
Catalog No. 50-972-635 Supplier Ace Glass, Inc. Supplier No. 593412
10 microliter syringe with 50mm 26 gauge removable needle for precise liquid delivery.