Learn More
Ace Glass, Inc. 50mm 25 microliter, 25 gauge, removable needle syringe
SDP
Catalog No. 50978618
Encompass
25 microliter syringe with 50mm 25gauge removable needle for precise liquid delivery.
Catalog No. 50-978-618 Supplier Ace Glass, Inc. Supplier No. 593414
25 microliter syringe with 50mm 25gauge removable needle for precise liquid delivery.