Learn More
Ace Glass, Inc. 50MM 4MM BORE ADAPTER 50MM 4MM BORE ADAPTER
SDP
Catalog No. 50978484
Encompass_Preferred
50mm adapter with drip tip for chemical type chromatography. Adapter has 4mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.
Catalog No. 50-978-484 Supplier Ace Glass, Inc. Supplier No. 583521
50mm adapter with drip tip for chemical type chromatography. Adapter has 4mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.