Learn More
Chemglass Life Sciences Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 50mm, Flange ID: 34.3mm, Shank OD: 38.1mm
SDP
Catalog No. CG14601
Encompass_Preferred
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 50mm, Flange ID: 34.3mm, Shank OD: 38.1mm.
Catalog No. CG14601 Supplier Chemglass Life Sciences Supplier No. CG14601
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 50mm, Flange ID: 34.3mm, Shank OD: 38.1mm.