Learn More
Ace Glass, Inc. 50MM CONNECTION ADAPTER,3 END
SDP
Catalog No. 50972224
Encompass_Preferred
50mm connecting adapter with #50 Ace-Threds at top, bottom, and side joints.
Catalog No. 50-972-224 Supplier Ace Glass, Inc. Supplier No. 582914
50mm connecting adapter with #50 Ace-Threds at top, bottom, and side joints.