Learn More
Azzota Corp 50mm Fluor Cuvette 17.5ml Qz
Catalog No. 5000887091
Encompass_Preferred
50mm Pathlength Standard Fluorimeter Cuvettes 45x12 5x52 5mm 17 5ml Quartz 190nm-2500nm Four polished windows
Catalog No. 50-008-87091 Supplier Azzota Corp Supplier No. Q208
50mm Pathlength Standard Fluorimeter Cuvettes 45x12 5x52 5mm 17 5ml Quartz 190nm-2500nm Four polished windows