Learn More
Azzota Corp 50mm Pathlength Standard Cuvettes - 17.5Ml Pathlength - 50mm Inside Width - 10mm Volume - 17.5Ml Dimensions - 45X12.5X52.5mm Cover And Round Bottom Optical Glass (340Nm-2500Nm)
Catalog No. 507532882
Encompass_Preferred
50mm Pathlength Standard Cuvettes - 17.5ml Pathlength - 50mm Inside width - 10mm Volume - 17.5ml Dimensions - 45x12.5x52.5mm Cover and round bottom Optical Glass (340nm-2500nm)
Catalog No. 50-753-2882 Supplier Azzota Corp Supplier No. G108
50mm Pathlength Standard Cuvettes - 17.5ml Pathlength - 50mm Inside width - 10mm Volume - 17.5ml Dimensions - 45x12.5x52.5mm Cover and round bottom Optical Glass (340nm-2500nm)