Learn More
Ace Glass, Inc. 50MM GRADED SEALS 50MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845550
Encompass
50mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-607