Learn More
Ace Glass, Inc. 50mm inner sublimation without O-ring
SDP
Catalog No. 50975880
Encompass_Preferred
50mm inner sublimation, part of ACE sublimation apparatus used for samples of approximately 25mL. Inner sublimation does not include o-ring.
Catalog No. 50-975-880 Supplier Ace Glass, Inc. Supplier No. 802206
50mm inner sublimation, part of ACE sublimation apparatus used for samples of approximately 25mL. Inner sublimation does not include o-ring.