Learn More
Teknova 50mM MES, 1M Sodium Chloride, pH 6.0, 1000mL, Endotoxin Tested, Sterile. 8 pack.
SDP
Catalog No. 502745918
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-274-5918 Supplier Teknova Supplier No. M50118PK
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.