Learn More
Teknova 50mM MES, pH 6.0, 1000mL, Endotoxin Tested, Sterile.
SDP
Catalog No. 502746035
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
This item is not returnable. View return policy
Catalog No. 50-274-6035 Supplier Teknova Supplier No. M9505
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.