Learn More
Ace Glass, Inc. 50mm adapter, nylon, polyethylene packing support, Luer slip bottom drip with flow regulator valve
SDP
Supplier:  Ace Glass, Inc. 583723
Encompass_Preferred
80mm bottom-drip, Luer-Lok, nylon adapter for use with all #80 Ace- Thred chromatography columns. Bore is 1mm. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-972-242