Learn More
Ace Glass, Inc. 50mm outer sublimation with plug & O-ring
SDP
Catalog No. 50975881
Encompass_Preferred
50mm outer sublimation with plug and o-ring, part of ACE sublimation apparatus used for samples of approximately 25mL.
Catalog No. 50-975-881 Supplier Ace Glass, Inc. Supplier No. 802208
50mm outer sublimation with plug and o-ring, part of ACE sublimation apparatus used for samples of approximately 25mL.