Learn More
DC Scientific Glass Inc 50 mm Plastic Disposable Cells, Cost-Effective, Minimize Cross-Contamination, 50 Cells/Pack
SDP
Catalog No. NC2046838
Encompass_Preferred
These 50 mm Plastic Disposable Cells come in a convenient pack of 50 and are designed for general laboratory use. They provide a cost-effective solution with a precise 50 mm path length, reducing the risk of cross-contamination and eliminating the need for cleaning. Manufactured by DC Scientific Glass Inc, these cells are ideal for applications requiring high accuracy and convenience, distinguishing them from traditional glass options.
Catalog No. NC2046838 Supplier DC Scientific Glass Inc Supplier No. DCP352101
These 50 mm Plastic Disposable Cells come in a convenient pack of 50 and are designed for general laboratory use. They provide a cost-effective solution with a precise 50 mm path length, reducing the risk of cross-contamination and eliminating the need for cleaning. Manufactured by DC Scientific Glass Inc, these cells are ideal for applications requiring high accuracy and convenience, distinguishing them from traditional glass options.