Learn More
Ace Glass, Inc. 50mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.
SDP
Catalog No. 50974641
Encompass_Preferred
50mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-641 Supplier Ace Glass, Inc. Supplier No. 720542
50mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.