Learn More
Azzota Corp 50mm std cuvette square bottom
Catalog No. 5000887212
Encompass_Preferred
50mm Pathlength Optical Glass Standard Cuvettes range 340-2500nm square bottom
Catalog No. 50-008-87212 Supplier Azzota Corp Supplier No. G8
50mm Pathlength Optical Glass Standard Cuvettes range 340-2500nm square bottom