Learn More
Ace Glass, Inc. 50MM TEES 50MM TEES
SDP
Catalog No. 50974951
Encompass_Preferred
50mm tee with threaded top for use in air sampling applications. Tee¿s side and bottom tubes are reduced for use with ACE 7506 bushing.
Catalog No. 50-974-951 Supplier Ace Glass, Inc. Supplier No. 749508
50mm tee with threaded top for use in air sampling applications. Tee¿s side and bottom tubes are reduced for use with ACE 7506 bushing.