Learn More
Ace Glass, Inc. 50mm adapter, PTFE, polyethylene packing support, Luer slip bottom drip
SDP
Supplier:  Ace Glass, Inc. 583761
Encompass_Preferred
50mm bottom-drip, Luer-Lok, PTFE adapter with flow regulator valve for use with all #50 Ace- Thred chromatography columns. Bore is adjustable from 0-1mm max. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-972-247