Learn More
Ace Glass, Inc. Coupling, #50 to #50 Ace-Thred™;, PTFE, front seal FETFE; O-rings
SDP
Supplier:  Ace Glass, Inc. 584152
Encompass_Preferred
50mm diameter thread (#50 Ace-Thred) PTFE coupling. For coupling threaded columns together, or coupling threaded columns to end fittings, with single o-ring seal for leak-tight engagement. Supplied with two FETFE o-rings.
Catalog No. 50-978-502