Learn More
Teknova 50mM Tris pH 7.6, 1mM EDTA pH 8.0, 0.5% Tween-20. 10mL Tubes, Sterile, RNAse Tested. 25 Pack
SDP
Catalog No. NC2541211
Encompass
50mM Tris pH 7.6, 1mM EDTA pH 8.0, 0.5% Tween-20. 10mL Tubes, Sterile, RNAse Tested. 25 Pack
Catalog No. NC2541211 Supplier Teknova Supplier No. T5594
50mM Tris pH 7.6, 1mM EDTA pH 8.0, 0.5% Tween-20. 10mL Tubes, Sterile, RNAse Tested. 25 Pack