Learn More
Teknova 50mM Tris, 16% Ethanol, pH 7.5. 1000mL. Endotoxin Tested, Sterile.
SDP
Catalog No. 502746031
Encompass
50mM Tris,16% (v/v) Ethanol
Catalog No. 50-274-6031 Supplier Teknova Supplier No. T5119
50mM Tris,16% (v/v) Ethanol