Learn More
Ace Glass, Inc. 50mm x 61cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 590225
Encompass_Preferred
50mm x 61cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring.
Catalog No. 50-972-560